Learn More
Medchemexpress LLC Neodiosmin | 38665-01-9 | MFCD30719837 | 100 MG

Supplier: Medchemexpress LLC HYN4122100MG
Neodiosmin is a flavone glycoside isolated from the leaves of Citrus aurantium. This compound has a molecular weight of 608.54 and a chemical formula of C28H32O15. It appears as a light yellow to yellow solid and is classified under flavonoids, specifically flavones, and phenols (polyphenols). This product is intended for research use only.
- Flavone glycoside
- Isolated from Citrus aurantium
- Molecular weight of 608.54
- Chemical formula of C28H32O15
- Appears as a light yellow to yellow solid
- Purity of 99.37%
- Classified as a flavonoid
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.