Learn More
Medchemexpress LLC Neodiosmin | 38665-01-9 | MFCD30719837 | 99.4% | 608.54 g·mol⁻¹ | C28H32O15 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN412210MG
Encompass_Preferred
Neodiosmin is a flavone glycoside isolated from the leaves of Citrus aurantium and provided as an analytical research standard for laboratory use.
  • Analytical standard suitable for research and analytical applications.
  • High purity (99.4%).
  • Molecular weight 608.54 g·mol⁻¹ and formula C28H32O15.
  • Available in small milligram pack sizes for precise dosing.
  • Not for use in humans; intended for laboratory research only.
Catalog No. 50-002-89797