Learn More
Miele, Inc. Neodisher LA 4x3kg Powder
Supplier:  Miele, Inc. 94000090
Encompass_Preferred
Neodisher LA 4x3kg PowderContains high proportions of emulsifiers and dispersing agent.5 % polycarboxylates, non-ionic surfactants15 -30 % phosphates
Catalog No. 50-171-3105