Learn More
Strem, An Ascensus Company CAS# 13477-89-9. 250g. Neodymium(III) chloride hexahydrate (99.9%-Nd) (REO). MFCD00149801
SDP
Supplier:  Strem, An Ascensus Company 936030250G
Encompass
CAS# 13477-89-9. 250g. Neodymium(III) chloride hexahydrate (99.9%-Nd) (REO). MFCD00149801. Molecular Weight: 250.60 (358.69). Molecular Formula: NdCl3∙6H2O. Color/form: purple xtl. Strem# 93-6030. http://www.strem.com/catalog/v/93-6030/
Catalog No. 50-901-15974