Learn More
TARGETMOL CHEMICALS INC NEOGAMBOGIC ACID 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T572410MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Neogambogic acid an active ingredient in garcinia can inhibit the growth of some solid tumors and result in an anticancer effect it may be responsible for the inhibition of proliferation of human breast cancer cell line MCF-7 cells. purity: 99%
Catalog No. 50-257-9737