Learn More
Alkali Scientific Neomycin Sulfate, 1 Kg
SDP
Supplier:  Alkali Scientific NEO002CP
Encompass_Preferred
Neomycin Sulfate, in a 1kg powder form, is a broad-spectrum aminoglycoside antibiotic used extensively in laboratory research to study bacterial infections and antimicrobial resistance. Neomycin works by binding to bacterial ribosomes, disrupting protein synthesis and causing cell death. It is effective against a variety of Gram-negative bacteria and is commonly used in research to test the susceptibility of bacterial strains to antibiotics. Neomycin Sulfate is particularly valuable for in vitro studies on bacterial resistance and in food safety microbiology, where it is used to identify antimicrobial-resistant pathogens.
Catalog No. 50-006-61445