Learn More
Chemglass Life Sciences Filter Adapter, Neoprene, Complete Set To Include 1ea Of The 7 Listed Sizes
SDP
Supplier:  Chemglass Life Sciences CG140101
Encompass_Preferred
Neoprene filter adapters are designed to fit between a filtering funnel and a flask. For use with Buchner funnels, Gooch crucibles, filter tubes, etc. Can be used individually or nested with adjacent sizes. Neoprene withstands autoclaving. Includes one each of the following sizes. Size 1 = Height: 21mm ID Top/Bottom (mm): 17x7 OD Top/Bottom (mm): 21x11 Wall thickness: 2mm Size 2 = Height: 21mm ID Top/Bottom (mm): 22x11 OD Top/Bottom (mm): 27x16 Wall thickness: 2.5mm Size 3 = Height: 25mm ID Top/Bottom (mm): 31x16 OD Top/Bottom (mm): 32x22 Wall thickness: 3mm Size 4 = Height: 29mm ID Top/Bottom (mm): 39 x 22 OD Top/Bottom (mm): 46 x 29 Wall thickness: 3.5mm Size 5 = Height: 35mm ID Top/Bottom (mm): 50 x 30 OD Top/Bottom (mm): 58 x 38 Wall thickness: 4mm Size 6 = Height: 40mm ID Top/Bottom (mm): 60 x 36 OD Top/Bottom (mm): 69 x 45 Wall thickness: 4.5mm Size 7 = Height: 45mm ID Top/Bottom (mm): 75 x 46 OD Top/Bottom (mm): 86 x 57 Wall thickness: 5.5mm
Catalog No. CG140101