Learn More
Medchemexpress LLC HY-U00435 5mg Medchemexpress, Neoseptin 3 CAS:1622863-21-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYU004355MG
Encompass_Preferred
Medchemexpress, HY-U00435 5mg Neoseptin 3 CAS:1622863-21-1 Neoseptin 3 is a Toll-like receptor 4/myeloid differentiation factor 2 (mTLR4/MD-2) agonist with an EC50 of 18.5 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0630