Learn More
Apexbio Technology LLC Neostigmine Bromide 114-80-7 10mM (in 1mL H2O)
SDP
Supplier:  Apexbio Technology LLC B15945.1
Encompass_Preferred
Neostigmine Bromide (CAS 114-80-7) is a synthetic quaternary ammonium compound that acts as a reversible inhibitor of acetylcholinesterase (AChE) By blocking AChE activity neostigmine bromide prolongs the action of acetylcholine at neuromuscular junctions and cholinergic synapses thereby enhancing muscarinic and nicotinic receptor-mediated effects This compound is frequently studied in contexts involving neuromuscular function and its use extends to research on disorders such as myasthenia gravis postoperative urinary retention paralytic ileus and primary open-angle glaucoma Additionally it is investigated as an adjuvant for local anesthetics and opioids in pain management following gynecological or abdominal surgery
Catalog No. 50-000-90551