Learn More
Selleck Chemical LLC Neostigmine Bromide S2490-50mg
SDP
Supplier:  Selleck Chemical LLC S249050MG
Encompass_Preferred
Neostigmine Bromide is a reversible acetylcholinesterase inhibitor
Catalog No. 50-311-1745