Learn More
Medchemexpress LLC Neostigmine methyl sulfate | 51-60-5 | 98.9% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB1206R5MG
Encompass_Preferred
Neostigmine methyl sulfate (Standard) is the analytical standard of Neostigmine (methyl sulfate). This product is intended for research and analytical applications. It is the grade of analytical standard, which is the reference standard supplied assay. It is commonly used in qualitative, quantitative and methodological research experiments.
  • Used for research and analytical applications
  • Reference standard supplied assay
  • Commonly used in HPLC, GC and MS experiments
Catalog No. 50-004-13602