Learn More
Apexbio Technology LLC Nepafenac 78281-72-8 100mg
SDP
Supplier:  Apexbio Technology LLC A2890100
Encompass_Preferred
Nepafenac (CAS 78281-72-8) is a small-molecule inhibitor targeting cyclooxygenase enzymes COX-1 and COX-2 It is designed to inhibit these enzymes thereby reducing prostaglandin E2 (PGE2) biosynthesis and modulating ocular inflammatory pathways Nepafenac exerts its biological activity primarily through COX-1 and COX-2 inhibition In cell-based studies nepafenac demonstrates inhibitory activity with IC50 values of approximately 250 nM for COX-1 and 150 nM for COX-2 In LPS-induced inflammation models topical application reduces retinal PGE2 concentrations Additionally it has been studied as an inhibitor of cellular proliferation in human uveal melanoma cell lines (SP6 5 92 1 OCM-1 MKT-BR UW-1) Based on these pharmacological properties nepafenac holds research potential in ophthalmologic studies involving inflammatory pathways and ocular malignancies
Catalog No. 50-000-95223