Learn More
Medchemexpress LLC Nepafenac-d5 | 1246814-53-8 | MFCD28139024 | 98.8% | 259.31 g/mol | C15H9D5N2O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY17357S10MG
Encompass_Preferred
Nepafenac-d5 is the deuterium-labeled (D5) isotopologue of nepafenac supplied as an analytical standard for use as an internal standard in quantitative mass spectrometry and method development. It is provided at high chemical purity and is intended for LC-MS/MS and GC-MS applications, pharmacokinetic studies, and analytical validation. CAS: 1246814-53-8.
  • Deuterium-labeled internal standard for quantitative mass spectrometry.
  • High chemical purity (98.8%).
  • Isotopically labeled as D5 for clear mass shift from the native analyte.
  • Suitable for LC-MS/MS and GC-MS method development and validation.
  • Supplied in small quantities appropriate for analytical use.
  • Soluble in dimethyl sulfoxide (DMSO) for standard preparation.
Catalog No. 50-002-95396