Learn More
Selleck Chemical LLC Nepafenac 10mg 78281-72-8 AHR 9434, AL 6515
SDP
Supplier:  Selleck Chemical LLC S125510MG
Encompass_Preferred
Nepafenac (AHR 9434, AL 6515) is a prodrug of amfenac that acts as an inhibitor of COX-1 and COX-2 activity, used in the treatment of pain and inflammation associated with cataract surgery. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-783-5