Learn More
Medchemexpress LLC HY-N2572 5mg Medchemexpress, Nepetin CAS:520-11-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN25725MG
Encompass_Preferred
Medchemexpress, HY-N2572 5mg Nepetin CAS:520-11-6 Nepetin (6-Methoxyluteolin) is a natural flavonoid isolated from Eupatorium ballotaefolium HBK with potent anti-inflammatory activities. Nepetin inhibits IL-6, IL-8 and MCP-1 secretion with IC50 values of 4.43 μM, 3.42 μM and 4.17 μM, respectively in ARPE-19 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2905