Learn More
Bellco Glass, Inc. Nephelo Flask, 300mL, 3Bfl 14x130 Sidearm, 38mm Neck
SDP
Supplier:  Bellco Glass, Inc. 258114133
Encompass_Preferred
Ideal for growing, storing and mixing of aerobic and anaerobic cultures. Has a threaded neck and sidearm with clean-out port. 3 baffles increase oxygenation of cultures. Supplied with 38mm and 18mm rubber lined black phenolic cap.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 300mlIncludes: 38mm & 18mm Rubber Lined Black Phenolic CapSidearm Dimensions: 14x130mmCase Quantity: 4
Catalog No. 50-194-5583