Learn More
Eagle Biosciences Inc Nephrolyx RUO Test Kit, Size: 76 Samples (Double Determination)
SDP
Supplier:  Eagle Biosciences Inc 001100RUO
Encompass
The Nephrolyx RUO Test Kit is intended for the true measurement of Glomular Filtration Rate (GFR) via Iohexol-Plasma-Clearance in serum. The Nephrolyx RUO Test Kit is for research use only and should not be used for diagnostic procedures.Size: 76 Samples (Double Determination)Sensitivity: 1 ug/mLMeasuring Range: 8 – 500 ug/mLCalculation Time: 6 hours
Catalog No. NC3628495