Learn More
Cayman Chemical Neplanocin A Inhibitors
SDP
Supplier:  Cayman Chemical 10584500 UG
Encompass_Preferred
Potently and irreversibly inactivates SAH hydrolase (Ki = 8.39 nM); has antitumor activity against mouse leukemia L1210 cells and broad-spectrum antiviral activity
Catalog No. 50-288-9461