Learn More
Selleck Chemical LLC Nepsilon-Acetyl-L-lysine
SDP
Supplier:  Selleck Chemical LLC S607525MG
Encompass_Preferred
Nepsilon-Acetyl-L-lysine is an R-chain N-acetylated amino acid used together with other lysine analogues to differentiate and characterized various aminoacylases and regulator 2 (Sir2) enzymes/sirtuins
Catalog No. 50-000-04325