Learn More
Medchemexpress LLC Nepsilon-Acetyl-L-lysine | 692-04-6 | 99.9% | 188.22 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY113426500MG
Encompass_Preferred
Nepsilon-Acetyl-L-lysine is an endogenous metabolite and an R-chain N-acetylated α amino acid. It is intended for research use only and not sold to patients.
  • Endogenous metabolite
  • R-chain N-acetylated alpha amino acid
  • Suitable for research use only
  • Associated with positive correlations with Akkermansia in cardiac fibrosis mice
Catalog No. 50-004-35562