Learn More
Medchemexpress LLC Nepsilon-Acetyl-L-lysine | 692-04-6 | 99.97% | 188.22 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1134261G
Encompass_Preferred
Nepsilon-Acetyl-L-lysine is an endogenous metabolite and an R-chain N-acetylated α amino acid. It is intended for research use only and not for sale to patients.
  • Endogenous metabolite
  • R-chain N-acetylated α amino acid
  • Used in research
Catalog No. 50-004-36705