Learn More
Andwin Scientific NEPTUNE 200UL TIPS 9600/CS
SDP
Supplier:  Andwin Scientific 89140900
Encompass_Preferred
The ESP Pipet Tip Reloading Systems allows reuse of existing trays, making this product environmentally friendly. It also reduces plastic waste by 90% with its extraordinary transfer system. It can reload trays within a minute and a half or 90 seconds. The S3 tips have universal fit and low retention. They have a volume capacity of 200 L and are nonsterile. The tips are made with virgin polypropylene and have thin wall that ensures an optimal seal. This is sold in a case of 9600.
Catalog No. NC0575394