Learn More
Medchemexpress LLC Neratinib | 698387-09-6 | MFCD09752958 | 99.9% | 557.0 g/mol | 1 ML
SDP
Supplier:  Medchemexpress LLC HY3272110MM/1ML
Encompass_Preferred
Neratinib (HKI-272) is an orally available, irreversible, highly selective HER2 and EGFR inhibitor with IC50s of 59 nM and 92 nM, respectively.
  • Inhibits proliferation of cell lines with high HER-2 levels.
  • Less active in cell lines not expressing HER-2 or EGFR.
  • Arrests cell cycle at G1-S phase.
  • Causes inhibition of MAPK and Akt phosphorylation, down-regulation of cyclin D1 levels, and induction of p27.
  • Shows anticancer activities against cancer cells expressing high levels of HER-2 or EGFR in vivo.
  • Reduces tumor growth in a dose-dependent manner in xenografts.
  • Inhibits phosphorylation of HER-2 in xenografts.
Catalog No. 50-004-04046