Learn More
Selleck Chemical LLC Neratinib (HKI-272) 25mg 698387-09-6
SDP
Supplier:  Selleck Chemical LLC S215025MG
Encompass_Preferred
Neratinib (HKI-272) is a highly selective HER2 and EGFR inhibitor with IC50 of 59 nM and 92 nM in cell-free assays; weakly inhibits KDR and Src, no significant inhibition to Akt, CDK1/2/4, IKK-2, MK-2, PDK1, c-Raf and c-Met. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-665-2