Learn More
Medchemexpress LLC Neratinib (Standard) | 698387-09-6 | 99.3% | 557.04 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY32721R
Encompass_Preferred
Neratinib (Standard) is the analytical standard of Neratinib, intended for research and analytical applications. Neratinib (HKI-272) is an orally available, irreversible, highly selective HER2 and EGFR inhibitor with IC50s of 59 nM and 92 nM, respectively.
  • Analytical standard for Neratinib
  • Used for research and analytical applications
  • Orally available
  • Irreversible
  • Highly selective HER2 and EGFR inhibitor
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS
Catalog No. 50-004-03366