Learn More
Medchemexpress LLC Neridronate | 79778-41-9 | 98.0% | 25 MG

Supplier: Medchemexpress LLC HY11944325MG
Neridronate is an aminobisphosphonate research chemical used to study bone metabolism and bisphosphonate pharmacology. Supplied as a white to off-white solid, it is intended for laboratory research use only and not for clinical or veterinary applications.
- Purity 98.0%
- Pack size 25 MG
- Molecular formula C6H17NO7P2
- Molecular weight 277.15 g/mol
- Appearance white to off-white solid
- CAS number 79778-41-9
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.