Learn More
Medchemexpress LLC Nerigliatin | 1245603-92-2 | 99.9% | 432.43 g/mol | C22H20N6O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10832825MG
Encompass_Preferred
Nerigliatin is an orally active glucokinase activator provided as a research/analytical standard. It is reported to activate glucokinase (EC50 ~154.4 μM), has CAS 1245603-92-2, molecular formula C22H20N6O4, and molecular weight 432.43 g/mol. The product is offered as solid packs and as a 10 mM solution in DMSO for laboratory use, with reported high purity.
Catalog No. 50-002-42221