Learn More
Medchemexpress LLC Neriifolin | 466-07-9 | MFCD00869436 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN844110MM/1ML
Encompass_Preferred
Neriifolin is a CNS-penetrating cardiac glycoside and an inhibitor of Na+, K+-ATPase. It targets beclin 1, which inhibits the formation of LC3-associated phagosomes and helps to ameliorate the development of experimental autoimmune encephalomyelitis (EAE). Additionally, Neriifolin can induce cell cycle arrest and apoptosis in human hepatocellular carcinoma HepG2 cells.
  • Targets Atg8/LC3, Na+/K+ ATPase, Apoptosis, Beclin1
  • Used in cardiovascular disease research
  • Used in inflammation/immunology research
  • Used in cancer research
  • Inhibits the formation of LC3-associated phagosomes
  • Ameliorates experimental autoimmune encephalomyelitis development
  • Induces cell cycle arrest and apoptosis in human hepatocellular carcinoma HepG2 cells
Catalog No. 50-004-17407