Learn More
Medchemexpress LLC Nerolidol | 7212-44-4 | 222.37 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN194410MM/1ML
Encompass_Preferred
Nerolidol has multiple natural membrane activities, possessing anti-cancer, anti-inflammatory, antibacterial, and anti-insect activity. It suppresses parasitic activity, bloodsucking diseases, bloodworm diseases, and other diseases. It protects cells from lipid and protein damage, damage to DNA, and generally protects cells from damage.
  • Exhibits potent antioxidant properties, counterbalancing the effect of ROS by protecting cells against oxidative damage to lipids, proteins, and DNA.
  • Exhibits potent antimicrobial activity against *Staphylococcus aureus*.
  • Exhibits anti-biofilm activity against a number of pathogens.
  • Has a strong anti-fungal effect against *Microsporum gypseum*.
  • Shows neuroprotective effects in mice hippocampus against oxidative stress in neuronal cells.
Catalog No. 50-004-15433