Learn More
Medchemexpress LLC Nerolidol | 7212-44-4 | 222.37 | 5 G
SDP
Supplier:  Medchemexpress LLC HYN19445G
Encompass_Preferred
Nerolidol possesses multiple natural membrane activities, including anti-cancer, anti-inflammatory, antibacterial, and anti-insect properties. It suppresses parasitic activity, bloodsucking diseases, bloodworm diseases, and other conditions. Additionally, Nerolidol protects cells from lipid and protein damage, as well as DNA damage.
  • Potent antioxidant properties
  • Antimicrobial activity against *Staphylococcus aureus*
  • Anti-biofilm activity against various pathogens
  • Strong anti-fungal effect
  • Neuroprotective effects in mice hippocampus
Catalog No. 50-004-15434