Learn More
TARGETMOL CHEMICALS INC NESOSTEINE LITHIUM 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T33647L5MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Nesosteine Lithium is a novel mucoregulatory that inhibits Herxheimer microconvulsions in guinea pigs and inhibits ovalbumin-induced histamine release from sensitized trachea.Nesosteine Lithium protects against ovalbumin-induced bronchospasm but is ineffective against histamine- and acetylcholine-induced bronchospasm. Nesosteine Lithium protected against ovalbumin-induced bronchospasm but not histamine- and acetylcholine-induced bronchospasm.
Catalog No. 50-258-0295