Learn More
Thomas Scientific 1.0mL 96-Well Deep Well Plate, U-Bottom, Round, Sterile
SDP
Supplier:  Thomas Scientific 1162C07
Encompass
Nest Deepwell Plates are made from virgin polypropylene, and certified Dnase/Rnase and pyrogen free. Unique plate bottom and raised rim design, ideal for magnetic bead based nucleic acid extraction and other applications Available at 24 well, 48 well, 96 well format; U-bottom or V-bottom, Square well or Round well Temperature range from -80°C to +121°C; autoclavable Certified Rnase, Dnase & Pyrogen Free Maximum centrifuge speed 4000 x g Withstands Organic Solvents like DMSO Low-Retention
Catalog No. NC1635394