Learn More
Medchemexpress LLC Nestoron | 7759-35-5 | 99.8% | 1 ML

Supplier: Medchemexpress LLC HY1307110MM/1ML
Nestoron (ST-1435) is a 19-norprogesterone derivative exhibiting high affinity and selectivity for progesterone receptors. This highly selective and potent progestogen has potential application as a hormonal contraceptive.
- Exhibits high affinity and selectivity for progesterone receptors.
- Acts as a highly selective and potent progestogen.
- Can be used as a hormonal contraceptive.
- Molecular weight: 370.48.
- Chemical formula: C23H30O4.
- Appears as a white to off-white solid.
- Soluble in DMSO at 33.33 mg/mL (89.96 mM).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.