Learn More
Medchemexpress LLC Netarsudil | 1254032-66-0 | 99.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1279850MG
Encompass_Preferred
Netarsudil is a competitive inhibitor of Rho-associated protein kinases (ROCK I and ROCK II) and a reversible inhibitor of the norepinephrine transporter (NET). It reduces intraocular pressure by acting on these pathways, primarily used in research for ocular hypertension and primary open-angle glaucoma.
  • Inhibits Rho-associated protein kinases (ROCK I and ROCK II).
  • Reversibly inhibits norepinephrine transporter (NET).
  • Reduces intraocular pressure by increasing aqueous humor outflow.
  • Used in the study of ocular hypertension.
  • Used in the study of primary open-angle glaucoma.
Catalog No. 50-003-42158