Learn More
Selleck Chemical LLC Netarsudil (AR-13324) 2HCl 2mg 1253952-02-1
SDP
Supplier:  Selleck Chemical LLC S82262MG
Encompass_Preferred
Netarsudil (a.k.a. AR-13324) is ROCK inhibitor with Ki value of 0.2-10.3 nM. It is currently in clinical trials for the treatment of glaucoma and ocular hypertension. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5817