Learn More
Medchemexpress LLC Netilmicin (sulfate) | 56391-57-2 | 98.0% | C21H41N5O7.5/2H2SO4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYA008610MM/1ML
Encompass_Preferred
Netilmicin sulfate is a broad-spectrum semisynthetic aminoglycoside antibiotic, effective against a variety of gram-negative and aminoglycoside-resistant bacterial strains. It is typically supplied as a white to off-white solid. This product is for research use only and not for medical applications; it is classified as harmful if swallowed or inhaled, and can cause skin/eye irritation.
  • Broad-spectrum semisynthetic aminoglycoside antibiotic
  • Antibacterial activity against gram-negative and resistant strains
  • Effective against *Escherichia coli* and *Pseudomonas aeruginosa*
  • Suitable for *in vitro* and *in vivo* research
  • Store at 4°C, sealed, away from moisture
  • For research use only
Catalog No. 50-004-32106