Learn More
Electron Microscopy Sciences Netwell™ Reagent Tray White
SDP
Supplier:  Electron Microscopy Sciences 6473210
Encompass_Preferred
White polystyrene reagent tray for Netwell Tissue Processing System, holds 150ml of solution.
Catalog No. 50-293-73