Learn More
ATCC Neural Progenitor Cells Derived from ATCC-BYS0112 Normal; Human
Supplier:  ATCC ACS5004
Encompass
These normal neural progenitor cells are derived from ATCC-BYS0112 Human [Non-Hispanic Caucasian Male] Induced Pluripotent Stem (IPS) Cells (ATCC ACS-1026) and can be used in neuronal differentiation and drug screening.
Catalog No. 50-238-3764