Learn More
Sigma Aldrich Fine Chemicals Biosciences Neuraminidase from Vibrio cholerae, 2 units
Supplier:  Sigma Aldrich Fine Chemicals Biosciences N78852UN
Encompass
Neuraminidase enzymes are glycoside hydrolase enzymes that catalyze hydrolysis of terminal sialic acid residues. The most well-known are the viral nearamidases, which promote influenza virus release.
Catalog No. NC1431568