Learn More
Medchemexpress LLC Neuraminidase-IN-1 | 2379438-80-7 | 98.9% | 317.25 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13733450MG
Encompass_Preferred
Neuraminidase-IN-1 is a neuraminidase inhibitor with an IC50 of 0.21 μM. It exhibits excellent activity against the H1N1 influenza virus.
  • Neuraminidase inhibitor
  • Excellent activity against H1N1 influenza virus
  • Good inhibitory effect on A/WSN/33H1N1 virus strains
  • Low toxicity and good druggability
Catalog No. 50-003-50281