Learn More
Medchemexpress LLC Neuraminidase-IN-1 | 2379438-80-7 | 98.9% | C14H11N3O6 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13733410MG
Encompass_Preferred
Neuraminidase-IN-1 is a small-molecule neuraminidase inhibitor reported to inhibit neuraminidase with an IC50 of 0.21 μM and to exhibit activity against H1N1 influenza virus strains. It is supplied as a high-purity research reagent for biochemical assays and antiviral screening, accompanied by a datasheet and safety data sheet.
  • Inhibits neuraminidase with an IC50 of 0.21 μM.
  • Displays activity against H1N1 influenza virus strains.
  • High purity (98.9%) suitable for research use.
  • Provided with structural data and supporting documentation (datasheet, SDS).
  • Available in multiple milligram-scale pack sizes for experimental flexibility.
Catalog No. 50-002-61372