Learn More
ACROBiosystems Neuraminidase NA 100ug
Supplier:  ACROBiosystems NEEV524C100UG
Encompass_Preferred
Influenza A [A/Victoria/4897/2022(H1N1)] Neuraminidase (NA) Protein His Tag (NEE-V524c) is expressed from human 293 cells (HEK293). It contains AA Val 80 - Asn 469 (Accession # EPI_ISL_17102775 GISAID).
Catalog No. 50-253-4369