Learn More
ACROBiosystems Neuraminidase NA 100ug
Supplier:  ACROBiosystems NEEV524M100UG
Encompass_Preferred
Influenza A [Wisconsin/67/2022(H1N1)] Neuraminidase (NA) Protein His Tag (NEE-V524m) is expressed from human 293 cells (HEK293). It contains AA Ser 31 - Asn 469 (Accession # EPI_ISL_15928538 GISAID).
Catalog No. 50-253-4361