Learn More
STEMCELL Technologies NeuroCult™ SM1 Without Insulin, Size: 10 mL
SDP
Supplier:  STEMCELL Technologies 05733
Encompass
NeuroCult™ SM1 Without Insulin is an optimized serum-free culture supplement, based on the published B27 formulation (Brewer et al.), with insulin removed. NeuroCult™ SM1 Without Insulin can be used in protocols where the effects of insulin are being studied, or where insulin may interfere with cellular function. In combination with a basal medium of choice, NeuroCult™ SM1 Without Insulin can be used for neural progenitor cell expansion, the study of neural stem and progenitor cell differentiation, and cardiomyocyte and hepatocyte differentiation protocols.
Catalog No. 50-197-4355