Learn More
Aviva Systems Biology Corp NEUROD2 Antibody - N-terminal region (ARP32037_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32037_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NEUROD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NEUROD2. Alternate names: NDRF, bHLHa1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rat
Catalog No. 50-014-54431