Learn More
Aves Labs, Inc Anti-NFH (Neurofilament Heavy Chain) Antibody
Supplier:  Aves Labs, Inc NFH
Encompass
Clonality: Polyclonal | Host Species: Chicken | Species Reactivity: Human, Mouse, Rat | Applications: ICC, IHC, WB | Protein Name / Synonyms: Neurofilament heavy polypeptide (NF-H) (200 kDa neurofilament protein) (Neurofilament triplet H protein)
Catalog No. NC1754349