Learn More
Medchemexpress LLC Neurofilament light polypeptide Antibody (YA1961) (PBS only) | 50 UL
SDP
Supplier:  Medchemexpress LLC HYP8221650UL
Encompass_Preferred
Neurofilament light polypeptide Antibody (YA1961) is a Rabbit-derived and non-conjugated IgG monoclonal antibody designed to target Neurofilament light polypeptide. This antibody is supplied in PBS, pH 7.4, and is affinity purified. Neurofilament light polypeptide (NEFL) is a crucial component of neurofilaments, forming structures with NEFM and NEFH to maintain neuronal caliber. It can also cooperate with neuronal intermediate filament proteins PRPH and INA to establish filamentous networks within neurons.
  • Rabbit-derived IgG monoclonal antibody
  • Non-conjugated
  • Targets Neurofilament light polypeptide
  • Supplied in PBS, pH 7.4
  • Affinity purified
  • Stored at -20°C for 1 year
  • Avoid repeated freeze/thaw cycles
  • Appears as a liquid
  • Shipped with blue ice
  • Supports neuronal caliber maintenance
  • Aids in establishing filamentous networks in neurons
Catalog No. 50-004-43903