Learn More
Cusabio Technology LLC Human Neurofilament protein L (NF-L) ELISA kit
Supplier:  Cusabio Technology LLC CSBE16094H
Encompass_Preferred
This Human NEFL ELISA Kit was designed for the quantitative measurement of Human NEFL protein in serum, plasma, tissue homogenates. It is a Sandwich ELISA kit, its detection range is 31.25 pg/mL-2000 pg/mL and the sensitivity is 7.8 pg/mL. One kit contains 96 wells.
Catalog No. NC2952969