Learn More
STEMCELL Technologies NeuroFluor™ NeuO, Size: 0.1 mL
SDP
Supplier:  STEMCELL Technologies 01801
Encompass
NeuroFluor™ NeuO is a membrane-permeable fluorescent probe that selectively labels primary and pluripotent stem cell-derived neurons in live cultures (J C Er et al. Angew Chem Int Ed, 2015). Cells labeled with NeuroFluor™ NeuO can be visualized using fluorescent imaging. Labeling with this probe is non-permanent; it can be washed off, providing unlabeled, viable cells for downstream applications. For additional information, see References.
Catalog No. NC1363914
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov