Learn More
CPC Scientific H-Asp-Met-His-Asp-Phe-Phe-Val-Gly-Leu-Met-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific NEUP003B
Encompass
Neuromedin K belongs to the tachykinin family and may play an important role in the olfactory, gustatory, visceral, and neuroendocrine processing information. In addition, it is a potent bronchioconstrictor and has neruomodulatory roles in various brain functions.ONE-LETTER SEQUENCE: DMHDFFVGLM-NH2MOLECULAR FORMULA: C55H79N13O14S2MOLECULAR WEIGHT:1210.44STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [86933-75-7], RESEARCH AREA: NeuropeptidesREFERENCES: K. Kangawa et al.; BBRC, 114, 533 (1983); S. Kimura et al., Proc. Japan Academy, 59, 101 (1983)
Catalog No. 50-210-9885